Safety gas tubing
供应商: DEUTSCH NEUMANN
3608050 3608075 3608100 3608150 3608060 3608125 3608200 3608300
DENE3608050EA 0 CNY
DENE3608050 DENE3608075 DENE3608100 DENE3608150 DENE3608060 DENE3608125 DENE3608200 DENE3608300
Safety gas tubing
胶管 Transfer Tubing
Red rubber, spiraling stainless steel wire core.
  • Flexible and resistant to buckling
  • Cut, tear and temperature resistant
  • Complies with DIN 12898
DVGW-Reg.-N°. G84e 100 D&N
Certifications: DVGW-Reg.-N°. G84e 100 D&N