Knitted gloves, Edge® 76-100
供应商: Ansell
76-100/9 76-100/7
ANSE76-100/9EA 0 CNY
ANSE76-100/9 ANSE76-100/7
Knitted gloves, Edge® 76-100
手套
Ambidextrous, light and stretchy gloves made from cotton.
  • Can be worn on their own or as liners
  • No seams to chafe or split
  • Stringknit construction allows good airflow to the hand
  • Washable at 40 °C, will not shrink, stiffen or lose their shape
Length: 220 mm
Colour: Natural
EN 388 (002X), Cat. II
Certifications: EN 388:2016 (002XA), Cat. II