Chemical resistant gloves, UltraNeo 382
供应商: MAPA
34382049 34382036 34382037 34382038 34382040
MAPA34382049EA 0 CNY
MAPA34382049 MAPA34382036 MAPA34382037 MAPA34382038 MAPA34382040
Chemical resistant gloves, UltraNeo 382
手套
These anatomically designed, liquid-proof gloves are made of blue neoprene over a cotton knit lining. They feature curved fingers, a contoured palm and a non slip finish on the palm and fingers area.
  • Two gloves in one for safer chemical protection
  • Raised pattern provides optimum grip
  • High comfort and thermal insulation thanks to the cotton lining
  • Good mechanical resistance
Length: 355 mm
Thickness: 0,9 mm
Colour: Blue
EN 374 (ACKL), EN 388 (2121), EN 407 (X1XXXX), Cat. III; AQL 1.5
Certifications: EN ISO 374-1 Type A (ALMNST), EN 374-5, EN 388:2016 (2121X), EN 407 (X1XXXX), Cat. III; AQL 1.5