Chemical resistant gloves, AlphaTec® 15-554
供应商: Ansell
15-554/9 15-554/10
ANSE15-554/9EA 0 CNY
ANSE15-554/9 ANSE15-554/10
Chemical resistant gloves, AlphaTec® 15-554
手套
These comfortable anatomical gloves with pre-curved fingers and winged thumb are made from a PVA compound, which remains inert in the strongest organic, aromatic and chlorinated solvents.
  • Polyvinyl alcohol coating lasts 10 - 15 times longer than any other coating
  • Soft two-piece knitted liner cushions the hand and absorbs perspiration
  • Virtually no absorption or extraction of ions
  • Exceptionally light and flexible
Length: 355 mm
Colour: Red
EN 374 (BCD), EN 388 (3121), EN 1149, Cat. III; AQL 1.0
Certifications: EN ISO 374-1:2016 Type A (BCDFIJ), EN ISO 374-5:2016, EN 388:2016 (3121A), Cat. III; AQL 1.0
Caution: Not for use in water or water-based solutions.