Cleanroom gloves, nitrile, AdvanTech 529
供应商: MAPA
34529000 34529006 34529008 34529009 34529007
MAPA34529000EA 0 CNY
MAPA34529000 MAPA34529006 MAPA34529008 MAPA34529009 MAPA34529007
Cleanroom gloves, nitrile, AdvanTech 529
手套
These ambidextrous, liquidproof gloves with beaded cuffs are made of nitrile.
  • Optimal thickness providing superior dexterity and ease of movement
  • 100% nitrile formulation eliminating protein allergy concerns
  • Lightly textured fingertips for an enhanced wet and dry grip
  • Long length to keep glove up on garment sleeve
The nitrile formulation provides superior mechanical resistance and splash protection against chemicals used in controlled environments. Free of pigments, powder, silicone or paraffin. Ideal for use in pharmaceutical industries, medical device manufacturing or microchip assembly.
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certifications: EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421, Cat. III; AQL 1,5.
Packaging: Packaged in Class 10/M2.5 compatible, IPA resistant cleanroom bags. Lot traceability on inner bags and cartons.