Solo Plus 995 Latex Gloves

供应商: MAPA
34995426 34995427 34995428 34995429
MAPA34995426EA 0 CNY
MAPA34995426 MAPA34995427 MAPA34995428 MAPA34995429
Solo Plus 995 Latex Gloves
手套
These multipurpose, ambidextrous gloves with beaded cuffs are made of natural latex.

  • Non sterile, chlorinated
  • Smooth with pebbled fingertips
  • Also suitable for common tasks in hospitals and clinics

Ideal for manufacturing of medicines, pharmaceutical preparation, research, analysis and handling of precision parts.

Length: 245 mm

Thickness: 0,10 mm

Colour: Green

EN 374 (Cat. III), AQL 1,5

Certifications: EN ISO 374-1 Type C, EN ISO 374-5; Cat. III; AQL 1,5
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR