Conical sleeves for vacuum filtration
供应商: DEUTSCH NEUMANN
2010181 2010011 2010021 2010031 2010041 2010051 2010061 2010071 2010081 2010022 2010032 2010042 2010182 2010052 2010012 2010062 2010072 2010082
DENE2010181EA 0 CNY
DENE2010181 DENE2010011 DENE2010021 DENE2010031 DENE2010041 DENE2010051 DENE2010061 DENE2010071 DENE2010081 DENE2010022 DENE2010032 DENE2010042 DENE2010182 DENE2010052 DENE2010012 DENE2010062 DENE2010072 DENE2010082
Conical sleeves for vacuum filtration
底座 Support Sleeves
Rubber, autoclavable.
Conical, for use with filtering flasks and filtering funnels.