DURAN® Funnels with short stem, ribbed
供应商: DWK LIFE SCIENCES
213523802 213524104 213524601 213525709 213526105
SCOT213523802EA 0 CNY
SCOT213523802 SCOT213524104 SCOT213524601 SCOT213525709 SCOT213526105
DURAN® Funnels with short stem, ribbed
漏斗 过滤漏斗 Filter Funnels
DURAN®, borosilicate glass 3.3, clear, ribbed walls.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration. Cone angle 60°. Funnel lip is ground and fire-polished.