Funnels, powder, Nalgene®
供应商: Thermo Fisher Scientific
4252-0065 4252-0080 4252-0150 4252-0100
NALG4252-0065EA 0 CNY
NALG4252-0065 NALG4252-0080 NALG4252-0150 NALG4252-0100
Funnels, powder, Nalgene®
漏斗 粉末漏斗
PPCO, translucent.
  • Very good chemical resistance
  • Outer ribs prevent air from being trapped
  • Autoclavable
For transferring powdery materials. Parallel stem prevents blockages.
This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.