Funnels with short stem, DURAN®
供应商: DWK LIFE SCIENCES
213512308 213513304 213513801 213514103 213514609 213515108 213515708 213515905 213516104 213516901 213512805
SCOT213512308EA 0 CNY
SCOT213512308 SCOT213513304 SCOT213513801 SCOT213514103 SCOT213514609 SCOT213515108 SCOT213515708 SCOT213515905 SCOT213516104 SCOT213516901 SCOT213512805
Funnels with short stem, DURAN®
漏斗 过滤漏斗 Filter Funnels
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.
DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119)
Certifications: DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119).