Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
供应商: BRAND
36401 36402 36403 36404 36406 36407 36408
BRND36401EA 0 CNY
BRND36401 BRND36402 BRND36403 BRND36404 BRND36406 BRND36407 BRND36408
Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
烧瓶 容量瓶
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
*Wide neck
Certifications: DIN EN ISO 1042
订购须知 Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.