Culture bottles and culture flasks, DURAN®
供应商: DWK LIFE SCIENCES
214212402 214213201 217712401 217713209 217713603 217714402 217715407 217716309 217713903
SCOT214212402EA 0 CNY
SCOT214212402 SCOT214213201 SCOT217712401 SCOT217713209 SCOT217713603 SCOT217714402 SCOT217715407 SCOT217716309 SCOT217713903
Culture bottles and culture flasks, DURAN®
烧瓶 培养瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Culture bottles and culture flasks (erlenmeyer shape) with straight rim, suitable for either kapsenberg caps or metal caps (available separately).