Erlenmeyer flasks, narrow or wide neck, with beaded rim, DURAN®
供应商: DWK LIFE SCIENCES
212161407 212161707 212162403 212163202 212163605 212163905 212164404 212165409 212166302 212166808 212167307 212261405 212261705 212262401 212263209 212263603 212263903 212264402 212265407 212266309 219902702 212162806 212165306
SCOT212161407EA 0 CNY
SCOT212161407 SCOT212161707 SCOT212162403 SCOT212163202 SCOT212163605 SCOT212163905 SCOT212164404 SCOT212165409 SCOT212166302 SCOT212166808 SCOT212167307 SCOT212261405 SCOT212261705 SCOT212262401 SCOT212263209 SCOT212263603 SCOT212263903 SCOT212264402 SCOT212265407 SCOT212266309 SCOT219902702 SCOT212162806 SCOT212165306
Erlenmeyer flasks, narrow or wide neck, with beaded rim, DURAN®
烧瓶 三角烧瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.
With printed Retrace Code (batch identification); 214-1102 without Retrace Code.
*Non DIN ISO size
DIN ISO 24450 wide mouth (except 214-1152 and 214-1168)
Certifications: ISO 1773 narrow mouth
DIN ISO 24450 wide mouth