Erlenmeyer flasks, disposable, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
4112-0250 4112-2000 4112-0125 4112-1000
NALG4112-0250EA 0 CNY
NALG4112-0250 NALG4112-2000 NALG4112-0125 NALG4112-1000
Erlenmeyer flasks, disposable, with screw cap, Nalgene®
烧瓶 三角烧瓶
PETG, transparent, with HDPE screw cap.
  • Leakproof HDPE screw cap closure can be opened partially for venting by twisting a quarter turn
  • Packaged individually for easy storage and handling
  • Reduced risk of cross-contamination
Sterile, disposable Erlenmeyer flasks reduce the risk of cross-contamination. Ideal for shakers and suspension cultures, and for creating or storing media. Raised graduations for approximate reading of the contents.