Erlenmeyer flasks, wide neck, with screw caps, Nalgene®
供应商: Thermo Fisher Scientific
4109-0125 4109-0250 4109-0500 4109-1000
NALG4109-0125EA 0 CNY
NALG4109-0125 NALG4109-0250 NALG4109-0500 NALG4109-1000
Erlenmeyer flasks, wide neck, with screw caps, Nalgene®
烧瓶 三角烧瓶
PMP (TPX), transparent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 150 °C (up to 170 °C for short periods), autoclavable
  • Good microwave permeability
Ideal for titrating and for preparing and storing nutrient media.