Venting filters, Acro® 50
供应商: CYTIVA PALL
4251 4256 4258 4400 4401 4250 4003
PALL4251EA 0 CNY
PALL4251 PALL4256 PALL4258 PALL4400 PALL4401 PALL4250 PALL4003
Venting filters, Acro® 50
过滤器 排气过滤器
Reusable filters with superior performance for critical applications, ideal for use with bioreactors and fermenters and for filtration of aggressive solvents.
Membrane: PTFE on a hydrophobic PP support; housing: PP, autoclavable