Bottle top tilt measures
供应商: KARTELL
1475 1476 1477 1480 1481 1482
KART1475EA 0 CNY
KART1475 KART1476 KART1477 KART1480 KART1481 KART1482
Bottle top tilt measures
瓶顶倾斜检测
PE (bottle and tube), PMP (measuring cup).
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released only the desired volume remains
The 25 /50 ml measuring cup slides up and down the tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
Colour: Transparent /Red