Petri dish carrying rack, Poxygrid®, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
H189780000 H189790000
BELAH189780000EA 0 CNY
BELAH189780000 BELAH189790000
Petri dish carrying rack, Poxygrid®, SP Bel-Art
架子 培养皿分配器
Epoxy coated steel wire rack.
  • Easily loaded and emptied from the top
  • For plastic Petri dishes Ø 90 and 100 mm (not suitable for glass Petri dishes)
W×D×H: 108×108×356 mm (1 stack, for 20 dishes)
W×D×H: 343×105×305 mm (3 stacks, for 42 dishes)
W×D×H: 343×105×305 mm (3 stacks, for 42 dishes)