Petri dishes, Duroplan®, DURAN®
供应商: DWK LIFE SCIENCES
217554101 217554307 217554607 217554804 217555106 217555303
SCOT217554101EA 0 CNY
SCOT217554101 SCOT217554307 SCOT217554607 SCOT217554804 SCOT217555106 SCOT217555303
Petri dishes, Duroplan®, DURAN®
培养皿 培养皿
DURAN®, borosilicate glass 3.3, clear. This plate is used in the pharmaceutical or food processing industry, in hospital environments for sampling surfaces such as skin, operating taalsobles and work surfaces.
  • Base and lid are flat inside and out, free from bubbles and streaks
  • Allow even distribution of culture media
  • Distortion-free transparency
  • Resistant against temperature and chemicals
DIN 13132
Certifications: DIN 13132