Instrument trays, with lid
供应商: BOCHEM
8710 8711 8712 8713 8714 8715 8716 8717
BOCH8710EA 0 CNY
BOCH8710 BOCH8711 BOCH8712 BOCH8713 BOCH8714 BOCH8715 BOCH8716 BOCH8717
Instrument trays, with lid
盘子/皿/盆 仪表盘/皿
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance