Cell culture plates, Nunc™ Multidish
供应商: Thermo Fisher Scientific
150628 150687 140675 176740 140685 142485 142475 152640 167063 167064
NUNC150628EA 0 CNY
NUNC150628 NUNC150687 NUNC140675 NUNC176740 NUNC140685 NUNC142485 NUNC142475 NUNC152640 NUNC167063 NUNC167064
Cell culture plates, Nunc™ Multidish
微孔板 细胞培养板
PS, sterile, with lid. Useful in all areas of cell culture, including scale up and cloning.
  • Raised well rims lower the risk of cross-contamination
  • Flat bottom wells allow optimum optical quality
  • Nunclon™Δ certified surface treatment