DURAN® Screw Caps with Aperture
供应商: DWK LIFE SCIENCES
292270508 292270602 292270902 292271007 292270808
SCOT292270508EA 0 CNY
SCOT292270508 SCOT292270602 SCOT292270902 SCOT292271007 SCOT292270808
DURAN® Screw Caps with Aperture
外壳 瓶盖
PBT, red without seals.
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable
With central hole.