Allihn condensers with standard ground joints
供应商: LENZ LABORGLAS
05223211 05223314 05223414 05223413 05223213 05223212 05223214 05223311 05223312 05223313 05223514
LENZ05223211EA 0 CNY
LENZ05223211 LENZ05223314 LENZ05223414 LENZ05223413 LENZ05223213 LENZ05223212 LENZ05223214 LENZ05223311 LENZ05223312 LENZ05223313 LENZ05223514
Allihn condensers with standard ground joints
冷凝器
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With core and jacket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
201-1168 and 201-1678 according to DIN 12581
Certifications: 201-1168 and 201-1678 according to DIN 12581