Screw caps for laboratory bottles, DURAN®
供应商: DWK LIFE SCIENCES
292391907 292392809 293382884 292391307 293382802 293382868
SCOT292391907EA 0 CNY
SCOT292391907 SCOT292392809 SCOT293382884 SCOT292391307 SCOT293382802 SCOT293382868
Screw caps for laboratory bottles, DURAN®
外壳 瓶盖
PP, coloured, with lip seal.
  • For DIN GL threads
  • Heat resistant up to +140 °C
  • Autoclavable