VWR®, Clamps
供应商: VWR Collection
VWRF08294.430EA 0 CNY
VWRF08294.430 VWRF08307.468
VWR®, Clamps
夹具 通用夹具
Stainless steel. Easy to use, robust clamps.
  • Independently adjustable jaws, jaw height 16 and 25 mm
  • With two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other