VWR®, Flash Chromatography Cartridges
供应商: VWR Collection
VWRI554-0265EA 0 CNY
VWRI554-0265
VWR®, Flash Chromatography Cartridges
色谱柱
Resistant to all normal phase solvents used.
  • Allow for liquid or solid sample loading
  • Axial compression of silica: no dead volume