Buckets, Nalgene®
供应商: Thermo Fisher Scientific
7012-0080 7012-0110 7012-0140
NALG7012-0080EA 0 CNY
NALG7012-0080 NALG7012-0110 NALG7012-0140
Buckets, Nalgene®
桶类
PP, white.
  • Graduated
  • Very good thermal and chemical resistance
  • Autoclavable
Universal multipurpose bucket without spout. Nylon coated handle provides comfortable sturdy grip.