Storage and Dispenser Boxes
供应商: BRAND
131900 131902
BRND131900EA 0 CNY
BRND131900 BRND131902
Storage and Dispenser Boxes
箱子 分配器盒
PMMA, white sides, transparent front.
  • Easy top loading and convenient dispensing through the front flap
  • Clear front allows contents to be checked easily
  • Cover and front flap limit contamination by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.