Bottles, wide neck, square, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2110-0002 2110-0006 2110-0008 2110-0016 2114-0002 2114-0006 2114-0008 2114-0016 2114-0032
NALG2110-0002EA 0 CNY
NALG2110-0002 NALG2110-0006 NALG2110-0008 NALG2110-0016 NALG2114-0002 NALG2114-0006 NALG2114-0008 NALG2114-0016 NALG2114-0032
Bottles, wide neck, square, with screw cap, Nalgene®
塑料瓶
HDPE, translucent or PPCO, translucent, with PP screw cap.
  • HDPE: Compatibility with most alcohols, bases and acids. PPCO: Very good chemical resistance
  • HDPE: Heat resistant up to +120 °C. PPCO: Heat resistant up to +121 °C, autoclavable
  • Leakproof
Ideal for storing and shipping samples. Wide mouth permits easy filling and cleaning. Square shape saves space.