Bottles, wide neck, with screw cap, for environmental samples, Nalgene®
供应商: Thermo Fisher Scientific
2189-0001 2189-0002 2189-0004 2189-0008 2189-0016 2189-0032
NALG2189-0001EA 0 CNY
NALG2189-0001 NALG2189-0002 NALG2189-0004 NALG2189-0008 NALG2189-0016 NALG2189-0032
Bottles, wide neck, with screw cap, for environmental samples, Nalgene®
塑料瓶
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Economical, break resistant disposable bottles for safe sample collection, transport and storage. Suitable for long-term storage in the absence of direct UV radiation.