Bottles, amber, wide neck, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2106-0002 2106-0008 2106-0016 2106-0032
NALG2106-0002EA 0 CNY
NALG2106-0002 NALG2106-0008 NALG2106-0016 NALG2106-0032
Bottles, amber, wide neck, with screw cap, Nalgene®
塑料瓶
HDPE, amber with amber PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof
Amber coloured to protect light-sensitive materials. Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.