Bottles, wide neck, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2105-0001 2105-0002 2105-0004 2105-0016
NALG2105-0001EA 0 CNY
NALG2105-0001 NALG2105-0002 NALG2105-0004 NALG2105-0016
Bottles, wide neck, with screw cap, Nalgene®
塑料瓶
PPCO, translucent with PP screw cap.
  • Very good chemical resistance
  • Heat resistant up to +121 °C, autoclavable
  • Leakproof
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.