Bottles, wide neck, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2104-0048 2104-0016 2104-0008 2104-0004 2104-0032
NALG2104-0048EA 0 CNY
NALG2104-0048 NALG2104-0016 NALG2104-0008 NALG2104-0004 NALG2104-0032
Bottles, wide neck, with screw cap, Nalgene®
塑料瓶
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof closure
Can be used anywhere in the laboratory and for sample collection. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.
These bottles are non sterile. Ideal for storing, shipping, and packaging liquids, uniform walls provide durability and resistance to splitting or punctures. Manufactured from non-toxic materials.