Wash bottles, wide neck, Unitary™, Nalgene®
供应商: Thermo Fisher Scientific
2402-0125 2402-0250 2402-0500 2402-0750 2402-1000
NALG2402-0125EA 0 CNY
NALG2402-0125 NALG2402-0250 NALG2402-0500 NALG2402-0750 NALG2402-1000
Wash bottles, wide neck, Unitary™, Nalgene®
洗瓶
LDPE, translucent, with PP screw cap.
  • Wide neck for quick, safe filling, leakproof closure
  • Formed from a single piece, outlet tube can no longer become contaminated or leak
  • Jet tip can be cut to increase flow rate
  • Good chemical resistance, heat resistant up to 80 °C