Wash bottles, narrow neck, Nalgene®
供应商: Thermo Fisher Scientific
2401-0125 2401-0500 2401-1000
NALG2401-0125EA 0 CNY
NALG2401-0125 NALG2401-0500 NALG2401-1000
Wash bottles, narrow neck, Nalgene®
洗瓶
LDPE, translucent with PP spray cap.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Closure and stem moulded in one piece for leakproof performance
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The jet tip can be cut to increase the flow rate.