Bottles, narrow neck, with standard ground stoppers, DURAN®
供应商: DWK LIFE SCIENCES
231652408 231653601 231654409 231655405 231681706 231682402 231684403 231685408
SCOT231652408EA 0 CNY
SCOT231652408 SCOT231653601 SCOT231654409 SCOT231655405 SCOT231681706 SCOT231682402 SCOT231684403 SCOT231685408
Bottles, narrow neck, with standard ground stoppers, DURAN®
玻璃瓶
Soda-lime glass, clear or brown, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and tough
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Certifications: DIN 12036, ISO 4796