Bottles, narrow neck, with standard ground stoppers, DURAN®
供应商: DWK LIFE SCIENCES
211651702 211652407 211653609 211654408 211655404 211656306 211657302 211658607 211659106 211681705 211682401 211683603 211684402 211685407 211686309 211687305 211688601 211689109
SCOT211651702EA 0 CNY
SCOT211651702 SCOT211652407 SCOT211653609 SCOT211654408 SCOT211655404 SCOT211656306 SCOT211657302 SCOT211658607 SCOT211659106 SCOT211681705 SCOT211682401 SCOT211683603 SCOT211684402 SCOT211685407 SCOT211686309 SCOT211687305 SCOT211688601 SCOT211689109
Bottles, narrow neck, with standard ground stoppers, DURAN®
玻璃瓶
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass type I according to USP, EP and JP
  • Amber glass meets USP <671> and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
ISO 4796-2.
With printed Retrace Code (batch identification).