Bottles, narrow neck, round, LDPE, with screw caps, Nalgene®
供应商: Thermo Fisher Scientific
2003-0004 2003-0032 2003-9025 2003-9050
NALG2003-0004EA 0 CNY
NALG2003-0004 NALG2003-0032 NALG2003-9025 NALG2003-9050
Bottles, narrow neck, round, LDPE, with screw caps, Nalgene®
塑料瓶
LDPE, translucent with PP screw cap.
  • Good chemical resistance, heat resistant up to +80 °C
  • Well suited for trace analysis
  • More flexible and impact resistant than HDPE bottles, leakproof
Ideal for storing and shipping liquids.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
环境认证 ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement
NAR Certificate of Retirement