Bottles, amber, narrow neck, round, HDPE, with screw caps, Nalgene®
供应商: Thermo Fisher Scientific
2004-0001 2004-9025 2004-9050 2004-9125
NALG2004-0001EA 0 CNY
NALG2004-0001 NALG2004-9025 NALG2004-9050 NALG2004-9125
Bottles, amber, narrow neck, round, HDPE, with screw caps, Nalgene®
塑料瓶
HDPE, amber with amber PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof
Amber to protect light-sensitive materials. Ideal for storing and shipping liquids.