Bottles, narrow or wide neck, square, with thread
供应商: DWK LIFE SCIENCES
238102456 238162453 238163655 238164454 238165459 238202405 238203607 238204406 238262402 238263604 238264403 238265408 238103658
SCOT238102456EA 0 CNY
SCOT238102456 SCOT238162453 SCOT238163655 SCOT238164454 SCOT238165459 SCOT238202405 SCOT238203607 SCOT238204406 SCOT238262402 SCOT238263604 SCOT238264403 SCOT238265408 SCOT238103658
Bottles, narrow or wide neck, square, with thread
玻璃瓶
Soda-lime glass, clear or amber, without screw caps.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Closures in various designs are available on request
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials.