Bottles, narrow neck, round, PC, with screw caps, Nalgene®
供应商: Thermo Fisher Scientific
DS2205-0010 DS2205-0020 DS2205-0210 DS2205-0250
NALGDS2205-0010EA 0 CNY
NALGDS2205-0010 NALGDS2205-0020 NALGDS2205-0210 NALGDS2205-0250
Bottles, narrow neck, round, PC, with screw caps, Nalgene®
塑料瓶
PC, transparent with PP screw cap.
  • Robust, impact resistant
  • Heat resistant up to 135 °C, autoclavable
  • Leakproof
Ideal for storing aqueous solutions and for short-term storage of culture media. The four and eight litre sizes have built-in shoulder loop for attaching an identification tag.