Carboys, wide neck, HDPE, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2212-0020 2212-0050 2211-0050 2211-0020
NALG2212-0020EA 0 CNY
NALG2212-0020 NALG2212-0050 NALG2211-0050 NALG2211-0020
Carboys, wide neck, HDPE, with screw cap, Nalgene®
carboy水瓶
HDPE, translucent or PP, translucent with white PP screw cap.
  • HDPE: Good chemical resistance/PP: Excellent chemical resistance
  • HDPE: Heat resistant up to 120 °C/PP: Heat resistant up to 135 °C, autoclavable
  • Leakproof
Sturdy, space saving design with indelible graduations in gallons and litres. Wide mouth for easy filling, emptying and cleaning. Particularly useful for handling powders and other solid substances in large quantities. Includes integral shoulder lugs and a sturdy, stainless steel handle.