Jerricans, fluorinated, HDPE, Nalgene®
供应商: Thermo Fisher Scientific
2242-0025 2242-0050
NALG2242-0025EA 0 CNY
NALG2242-0025 NALG2242-0050
Jerricans, fluorinated, HDPE, Nalgene®
油桶
HDPE, fluorinated with fluorinated PP screw cap.
  • Resistant to most acids, alkali compounds and aggressive organic solvents
  • Heat resistant from −100 to +120 °C
  • Stable sturdy design with moulded graduations in gallons and litres, leakproof
Improved retention properties and reduced solvent absorption and penetration thanks to fluorinated inner and outer surfaces. Fluorination improves the durability of the jerrican and reduces losses due to permeation. Large base and low centre of gravity provide stability. Pouring made easy with integral spout.