Bottles, narrow neck, round, LDPE or PP, with screw caps, Nalgene®
供应商: Thermo Fisher Scientific
2202-0005 2202-0010 2202-0020 2203-0010
NALG2202-0005EA 0 CNY
NALG2202-0005 NALG2202-0010 NALG2202-0020 NALG2203-0010
Bottles, narrow neck, round, LDPE or PP, with screw caps, Nalgene®
塑料瓶
LDPE, translucent or PPCO, translucent, with PP screw cap.
  • LDPE: Good chemical resistance/PPCO: Very good chemical resistance
  • LDPE: Heat resistant up to 80 °C/PPCO: Heat resistant up to 121 °C, autoclavable
  • Leakproof
Large volume bottles for shipping or storing chemicals, standard solutions, distilled water etc. The 4000 and 8000 ml sizes incorporate a loop for attaching a label.