Screw caps with silicone inserts, Top Works™ system, Nalgene®
供应商: Thermo Fisher Scientific
2135-3803 2135-5300 2135-5302 2135-5303 2135-8300 2135-8302 2135-8303
NALG2135-3803EA 0 CNY
NALG2135-3803 NALG2135-5300 NALG2135-5302 NALG2135-5303 NALG2135-8300 NALG2135-8302 NALG2135-8303
Screw caps with silicone inserts, Top Works™ system, Nalgene®
外壳 瓶盖
PP, white, with platinum cured silicone inserts.
  • Very good chemical resistance, heat resistant up to +135 °C, autoclavable
  • The Top Works™ system includes versions with solid, two or three port connections
  • Suitable for use in the aseptic transfer of liquids from most Nalgene® bottles and carboys
Platinum cured silicone tubing is fused through the stopper to form a single piece closure system.