VWR®, 一次性烧杯
供应商: VWR Collection
13915-985
VWRI611-1363EA 600 CNY
VWRI611-1363 VWRI611-1364 VWRI611-1365 VWRI611-1366 VWRI611-1367 VWRI611-1368 VWRU13915-985
VWR®, 一次性烧杯
烧杯
PS.
  • 适用于各种样品分配器
  • 进行火焰光度分析时,建议使用容量5ml和20ml的烧杯
Economical, multipurpose beaker for a very wide range of uses.