Beakers, low form, DURAN®
供应商: DWK LIFE SCIENCES
211060701 211060804 211061406 211061706 211062402 211062908 211063604 211064103 211064806 211065305 211065408 211066301 211066807 211067306 211068602
SCOT211060701EA 0 CNY
SCOT211060701 SCOT211060804 SCOT211061406 SCOT211061706 SCOT211062402 SCOT211062908 SCOT211063604 SCOT211064103 SCOT211064806 SCOT211065305 SCOT211065408 SCOT211066301 SCOT211066807 SCOT211067306 SCOT211068602
Beakers, low form, DURAN®
烧杯
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area (213-1101, 213-1102 not graduated and without Retrace Code).
With printed Retrace Code (batch identification).
ISO 3819 (except 213-1115).