生物危害垃圾袋
供应商: VWR Collection
14220-046 89129-468 89130-796 89129-470 89129-472 14220-066 14220-070 14220-052 14220-034 14220-048 14220-050
VWRI129-0025EA 3154 CNY
VWRI129-0025 VWRU14220-046 VWRU89129-468 VWRU89130-796 VWRU89129-470 VWRU89129-472 VWRU14220-066 VWRU14220-070 VWRU14220-052 VWRU14220-034 VWRU14220-048 VWRU14220-050 VWRI129-1564
生物危害垃圾袋
废物处理袋
这些结实耐用的袋子非常适合用来处置具有生物危害性、必须首先进行高温高压灭菌的废弃物。带金属丝封口,可扩大袋子并在冷却时通风
PP,厚度50微米,带“生物危害”标签
  • 可进行最高135℃的高温高压灭菌
  • 未高温高压灭菌前温度指示器文为白色,灭菌后为黑色。若出现“内容物已经过高温高压灭菌”,则表明成功完成高温高压灭菌
  • “生物危害”标签清晰可见,带相应的警告标志
The bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D 709-98.
Certifications: Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D 709-98.