Adapters with screw thread and standard gound joint cone
供应商: LENZ LABORGLAS
01311419 01311424 01311429 01311814 01311819 01311824 01312519 01312524 01311829 01312514 01312529 01311414
LENZ01311419EA 0 CNY
LENZ01311419 LENZ01311424 LENZ01311429 LENZ01311814 LENZ01311819 LENZ01311824 LENZ01312519 LENZ01312524 LENZ01311829 LENZ01312514 LENZ01312529 LENZ01311414
Adapters with screw thread and standard gound joint cone
适配器 带圆锥形接地接头的适配器
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground joint cone.
DIN 12257