o-Tolidine 0.03% with 0.2% potassium iodide in acetic acid 6% Reag. Ph. Eur. 1123001

供应商: VWR Chemicals

VWRC87971.260EA 1635 CNY
VWRC87971.260
o-Tolidine 0.03% with 0.2% potassium iodide in acetic acid 6% Reag. Ph. Eur. 1123001
o-Tolidine 0.03% with 0.2% potassium iodide in acetic acid 6%

订购须知 Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

分子式: [-C₆H₃(CH₃)-4-NH₂]₂
存储条件: Ambient
CAS号: 119-93-7
EINECS: 204-358-0

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR